Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATL43
Entrez Gene ID
830466
Gene Name
RING/U-box superfamily protein
Gene Aliases
AT5G05810, MJJ3.23, MJJ3_23
Chromosome Location
Chr.5: 1746548 - 1748327 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120663.3 NP_196200.3 1 - 1 1551 141

View Full Product Details