Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDX27
Entrez Gene ID
830522
Gene Name
nudix hydrolase homolog 27
Gene Aliases
AT5G06340, ATNUDX27, MHF15.14, MHF15_14, nudix hydrolase homolog 27
Chromosome Location
Chr.5: 1936339 - 1938134 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120717.5 NP_196252.1 2 - 3 406 74
GenBank mRNA AF372941.1 - 2 - 3 339 74
AY113163.1 - 2 - 3 278 74

View Full Product Details