Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BIM1
Entrez Gene ID
830708
Gene Name
basic helix-loop-helix (bHLH) DNA-binding superfamily protein
Gene Aliases
AT5G08130, T22D6.70, T22D6_70
Chromosome Location
Chr.5: 2606002 - 2610250 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125718.1 NP_001119190.1 4 - 5 1191 92
NM_001203328.2 NP_001190257.1 5 - 6 1138 92
NM_001203329.1 NP_001190258.1 5 - 6 1138 92
NM_001203330.2 NP_001190259.1 5 - 6 1150 92
NM_001342983.1 NP_001318507.1 5 - 6 974 92
NM_001342984.1 NP_001318508.1 5 - 6 974 92
NM_001342985.1 NP_001330723.1 5 - 6 1138 92
NM_001342986.1 NP_001330724.1 5 - 6 1150 92
GenBank mRNA AB493742.1 - 2 - 3 279 92
BT029780.1 - 2 - 3 279 92

View Full Product Details