Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BIM1
Entrez Gene ID
830708
Gene Name
basic helix-loop-helix (bHLH) DNA-binding superfamily protein
Gene Aliases
AT5G08130, T22D6.70, T22D6_70
Chromosome Location
Chr.5: 2606002 - 2610250 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125718.1 NP_001119190.1 8 - 9 1602 70
NM_001203328.2 NP_001190257.1 9 - 10 1549 70
NM_001203329.1 NP_001190258.1 9 - 10 1549 70
NM_001342983.1 NP_001318507.1 9 - 10 1385 70
NM_001342986.1 NP_001330724.1 9 - 10 1561 70
GenBank mRNA AB493742.1 - 6 - 7 690 70
BT029780.1 - 6 - 7 690 70

View Full Product Details