Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl35
Entrez Gene ID
830722
Gene Name
serine carboxypeptidase-like 35
Gene Aliases
AT5G08260, F8L15.17, serine carboxypeptidase-like 35
Chromosome Location
Chr.5: 2657054 - 2661437 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120909.4 NP_196443.1 7 - 8 1366 67
GenBank mRNA AF370244.1 - 7 - 8 1252 67
AY062969.1 - 7 - 8 1184 67

View Full Product Details