Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PE11
Entrez Gene ID
816699
Gene Name
pectinesterase 11
Gene Aliases
A. THALIANA PECTINESTERASE 11, AT2G21610, ATPE11, F2G1.12, F2G1_12, pectinesterase 11
Chromosome Location
Chr.2: 9245038 - 9247052 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335771.1 NP_001324138.1 1 - 2 294 91
NM_127732.2 NP_179755.1 1 - 2 267 91

View Full Product Details