Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CESA9
Entrez Gene ID
816713
Gene Name
cellulose synthase A9
Gene Aliases
AT2G21770, CESA09, F7D8.9, F7D8_9, cellulose synthase A9
Chromosome Location
Chr.2: 9284391 - 9289744 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127746.2 NP_179768.1 5 - 6 1221 79

View Full Product Details