Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNGC6
Entrez Gene ID
816931
Gene Name
cyclic nucleotide-gated channel 6
Gene Aliases
AT2G23980, ATCNGC6, CYCLIC NUCLEOTIDE GATED CHANNEL 6, T29E15.18, T29E15_18, cyclic nucleotide-gated channel 6
Chromosome Location
Chr.2: 10201104 - 10205422 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335878.1 NP_001318278.1 6 - 7 1405 151
NM_001335879.1 NP_001323882.1 4 - 5 1821 151
NM_001335880.1 NP_001323881.1 3 - 4 1776 151
NM_001335881.1 NP_001323885.1 5 - 6 1752 151
NM_001335882.1 NP_001323879.1 4 - 5 1707 151
NM_001335883.1 NP_001323880.1 3 - 4 1552 151
NM_001335884.1 NP_001323886.1 5 - 6 1474 151
NM_001335885.1 NP_001323884.1 4 - 5 1429 151
NM_001335886.1 NP_001323883.1 5 - 6 1360 151
NM_127960.3 NP_565560.1 5 - 6 1284 151
GenBank mRNA Y17914.1 - - 1184 151

View Full Product Details