Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD23B
Entrez Gene ID
844304
Gene Name
Rad23 UV excision repair protein family
Gene Aliases
AT1G79650, Arabidopsis thaliana aldehyde oxidase 1, AtAO1, F20B17.8, F20B17_8, RAD23, RADIATION SENSITIVE23B
Chromosome Location
Chr.1: 29971989 - 29975359 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198510.1 NP_001185439.1 11 - 12 1136 80
NM_106614.6 NP_565216.2 10 - 11 1124 80
NM_180651.3 NP_850982.1 10 - 11 1142 80
NM_202452.2 NP_974181.1 9 - 10 994 80
GenBank mRNA AY034912.1 - 10 - 11 1033 80
AY063103.1 - 10 - 11 897 80
AY088037.1 - 10 - 11 1054 80

View Full Product Details