Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD23B
Entrez Gene ID
844304
Gene Name
Rad23 UV excision repair protein family
Gene Aliases
AT1G79650, Arabidopsis thaliana aldehyde oxidase 1, AtAO1, F20B17.8, F20B17_8, RAD23, RADIATION SENSITIVE23B
Chromosome Location
Chr.1: 29971989 - 29975359 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198510.1 NP_001185439.1 1 - 2 211 116
NM_106614.6 NP_565216.2 1 - 2 289 116
NM_180651.3 NP_850982.1 1 - 2 289 116
NM_202452.2 NP_974181.1 1 - 2 201 116
GenBank mRNA AY034912.1 - 1 - 2 198 116
AY063103.1 - 1 - 2 62 116
AY088037.1 - 1 - 2 201 116

View Full Product Details