Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SGB1
Entrez Gene ID
844321
Gene Name
Major facilitator superfamily protein
Gene Aliases
AT1G79820, F20B17.24, F20B17_24, SUPPRESSOR OF G PROTEIN BETA1
Chromosome Location
Chr.1: 30022260 - 30027634 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036226.2 NP_001031303.1 12 - 13 1521 57
NM_001334924.1 NP_001323193.1 8 - 9 1003 57
NM_106631.5 NP_178100.3 13 - 14 1614 57
NM_180652.3 NP_850983.1 13 - 14 1408 57
GenBank mRNA AK317667.1 - 12 - 13 1335 57

View Full Product Details