Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SGB1
Entrez Gene ID
844321
Gene Name
Major facilitator superfamily protein
Gene Aliases
AT1G79820, F20B17.24, F20B17_24, SUPPRESSOR OF G PROTEIN BETA1
Chromosome Location
Chr.1: 30022260 - 30027634 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036226.2 NP_001031303.1 2 - 3 471 88
NM_001198511.1 NP_001185440.1 2 - 3 228 88
NM_106631.5 NP_178100.3 2 - 3 420 88
NM_180652.3 NP_850983.1 2 - 3 214 88
GenBank mRNA AK317667.1 - 1 - 2 141 88

View Full Product Details