Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SGB1
Entrez Gene ID
844321
Gene Name
Major facilitator superfamily protein
Gene Aliases
AT1G79820, F20B17.24, F20B17_24, SUPPRESSOR OF G PROTEIN BETA1
Chromosome Location
Chr.1: 30022260 - 30027634 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036226.2 NP_001031303.1 8 - 9 1093 68
NM_001198511.1 NP_001185440.1 8 - 9 850 68
NM_001334924.1 NP_001323193.1 4 - 5 575 68
NM_106631.5 NP_178100.3 8 - 9 1042 68
NM_180652.3 NP_850983.1 8 - 9 836 68
GenBank mRNA AK317667.1 - 7 - 8 763 68

View Full Product Details