Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNEE/G
Entrez Gene ID
814965
Gene Name
RNAse E/G-like protein
Gene Aliases
AT2G04270, RNAse E/G-like, RNE, RNase E, T23O15.10, T23O15_10
Chromosome Location
Chr.2: 1474679 - 1480586 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202581.2 NP_001189510.1 11 - 12 2731 90
NM_001335223.1 NP_001318196.1 11 - 12 2762 90
NM_001335224.1 NP_001325371.1 11 - 12 2728 90
NM_001335225.1 NP_001325372.1 10 - 11 2910 90
NM_001335226.1 NP_001325370.1 10 - 11 2907 90
NM_126460.4 NP_178508.2 8 - 9 1996 90
NM_180656.2 NP_850987.1 11 - 12 2808 90
NM_180657.2 NP_850988.1 11 - 12 2653 90
NM_180658.2 NP_850989.1 8 - 9 1996 90
GenBank mRNA AF450479.1 - 11 - 12 2808 90
AJ508604.1 - 9 - 10 2136 90
AY093213.1 - 8 - 9 1996 90
BT008793.1 - 7 - 8 1677 90

View Full Product Details