Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNEE/G
Entrez Gene ID
814965
Gene Name
RNAse E/G-like protein
Gene Aliases
AT2G04270, RNAse E/G-like, RNE, RNase E, T23O15.10, T23O15_10
Chromosome Location
Chr.2: 1474679 - 1480586 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202581.2 NP_001189510.1 3 - 4 593 124
NM_001335223.1 NP_001318196.1 3 - 4 613 124
NM_180656.2 NP_850987.1 3 - 4 670 124
GenBank mRNA AF450479.1 - 3 - 4 670 124

View Full Product Details