Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZIFL1
Entrez Gene ID
831220
Gene Name
zinc induced facilitator-like 1
Gene Aliases
AT5G13750, MXE10.2, MXE10_2, zinc induced facilitator-like 1
Chromosome Location
Chr.5: 4437804 - 4441701 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036800.2 NP_001031877.1 4 - 5 464 136
NM_001343292.1 NP_001318556.1 4 - 5 464 136
NM_121378.5 NP_568290.3 2 - 3 255 136
NM_180705.3 NP_851036.1 4 - 5 860 136
GenBank mRNA AY039542.1 - 4 - 5 409 136
AY062867.1 - 4 - 5 454 136
AY102150.1 - 4 - 5 346 136

View Full Product Details