Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYSD2
Entrez Gene ID
832872
Gene Name
cysteine synthase D2
Gene Aliases
AT5G28020, ATCYSD2, CYSTEINE SYNTHASE, CYSTEINE SYNTHASE D2, F15F15.90, F15F15_90, cysteine synthase D2
Chromosome Location
Chr.5: 10026152 - 10028722 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036879.2 NP_001031956.1 4 - 5 526 77
NM_001036880.3 NP_001031957.1 4 - 5 609 77
NM_001036881.1 NP_001031958.1 1 - 2 346 77
NM_001085159.1 NP_001078628.1 4 - 5 503 77
NM_122685.3 NP_198154.1 4 - 5 652 77
NM_180756.3 NP_851087.1 3 - 4 836 77
GenBank mRNA AB024283.1 - 3 - 4 825 77
AF424598.1 - 4 - 5 510 77
AY124831.1 - 3 - 4 467 77

View Full Product Details