Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHOT2
Entrez Gene ID
835926
Gene Name
phototropin 2
Gene Aliases
AT5G58140, AtPHOT2, K21L19.6, K21L19_6, NON PHOTOTROPIC HYPOCOTYL 1-LIKE, NPL1, phototropin 2
Chromosome Location
Chr.5: 23523825 - 23533111 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345288.1 NP_001318824.1 15 - 16 2510 129
NM_001345289.1 NP_001330355.1 14 - 15 2149 129
NM_001345290.1 NP_001330356.1 13 - 14 2499 129
NM_125199.2 NP_568874.2 14 - 15 1969 129
NM_180879.2 NP_851210.1 14 - 15 2326 129
NM_180880.2 NP_851211.1 14 - 15 2322 129
NM_180881.3 NP_851212.2 13 - 14 2107 129
GenBank mRNA AF053941.2 - 13 - 14 1864 129

View Full Product Details