Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDS
Entrez Gene ID
836181
Gene Name
4-hydroxy-3-methylbut-2-enyl diphosphate synthase
Gene Aliases
4-hydroxy-3-methylbut-2-enyl diphosphate synthase, AT5G60600, CHLOROPLAST BIOGENESIS 4, CLB4, CONSTITUTIVE SUBTILISIN 3, CSB3, GCPE, ISPG, MUP24.15, MUP24_15
Chromosome Location
Chr.5: 24358885 - 24363485 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125995.2 NP_001119467.1 16 - 17 2047 111
NM_001345427.1 NP_001332136.1 16 - 17 2238 111
NM_001345428.1 NP_001332137.1 16 - 17 2134 111
NM_125453.6 NP_200868.2 16 - 17 1963 111
NM_180902.5 NP_851233.1 16 - 17 2047 111
GenBank mRNA AF434673.1 - 16 - 17 1903 111

View Full Product Details