Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZWI
Entrez Gene ID
836723
Gene Name
kinesin-like calmodulin-binding protein (ZWICHEL)
Gene Aliases
AT5G65930, K14B20.10, K14B20_10, KCBP, KINESIN-LIKE CALMODULIN-BINDING PROTEIN, PKCBP, POTATO KINESIN-LIKE CALMODULIN-BINDING PROTEIN, ZWICHEL
Chromosome Location
Chr.5: 26369857 - 26376572 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203699.1 NP_001190628.1 17 - 18 3101 116
NM_125990.4 NP_569022.2 17 - 18 3208 116
NM_180945.2 NP_851276.1 17 - 18 3153 116

View Full Product Details