Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHD4
Entrez Gene ID
824309
Gene Name
Phosphoinositide phosphatase family protein
Gene Aliases
AT3G51460, ROOT HAIR DEFECTIVE 4, ROOT HAIR DEFECTIVE4
Chromosome Location
Chr.3: 19092518 - 19097437 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115005.5 NP_190714.2 2 - 3 520 61
GenBank mRNA AF266459.1 - 1 - 2 146 61
AF360267.1 - 2 - 3 240 61
AY040078.1 - 1 - 2 146 61
AY227250.1 - 1 - 2 146 61

View Full Product Details