Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMT3
Entrez Gene ID
824312
Gene Name
tonoplast monosaccharide transporter3
Gene Aliases
AT3G51490, tonoplast monosaccharide transporter3
Chromosome Location
Chr.3: 19104885 - 19107797 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203125.1 NP_001190054.1 3 - 4 1807 95
NM_001339519.1 NP_001327612.1 2 - 3 1893 95
NM_115008.1 NP_190717.1 3 - 4 1572 95
GenBank mRNA AJ532571.1 - 3 - 4 1572 95

View Full Product Details