Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SULTR3;1
Entrez Gene ID
824353
Gene Name
sulfate transporter 3;1
Gene Aliases
AST12, AT3G51895, SULFATE TRANSPORTER 1, sulfate transporter 3;1
Chromosome Location
Chr.3: 19251287 - 19255922 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339539.1 NP_001326166.1 2 - 3 795 66
NM_115049.5 NP_190758.2 2 - 3 667 66
GenBank mRNA AY099873.1 - 2 - 3 633 66

View Full Product Details