The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_115142.5 | NP_190850.2 | 3 - 4 | 1044 | 69 |
GenBank mRNA | AF492668.1 | - | 3 - 4 | 841 | 69 |