Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP71B30P
- Entrez Gene ID
-
824496
- Gene Name
- cytochrome P450, family 71, subfamily B, polypeptide 30 pseudogene
- Gene Aliases
-
''cytochrome P450, AT3G53290, cytochrome P450, family 71, polypeptide 30 pseudogene, polypeptide 30 pseudogene'', subfamily B
- Chromosome Location
-
Chr.3: 19758157 - 19759603 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115189.1 |
NP_190897.1 |
1 - 2 |
712 |
134 |
View Full Product Details