Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FIP37
Entrez Gene ID
824584
Gene Name
FKBP12 interacting protein 37
Gene Aliases
AT3G54170, ATFIP37, FKBP12 interacting protein 37
Chromosome Location
Chr.3: 20056664 - 20059583 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115277.5 NP_190985.1 10 - 11 965 127
GenBank mRNA AF084570.1 - 10 - 11 847 127
AY063940.1 - 10 - 11 872 127
AY113943.1 - 10 - 11 781 127

View Full Product Details