Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTN8
Entrez Gene ID
824632
Gene Name
Structural maintenance of chromosomes (SMC) family protein
Gene Aliases
AT3G54670, ATSMC1, SMC1, STRUCTURAL MAINTENANCE OF CHROMOSOMES 1, TITAN8
Chromosome Location
Chr.3: 20235506 - 20244046 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339686.1 NP_001319748.1 12 - 13 2777 65

View Full Product Details