Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXO70H1
Entrez Gene ID
824681
Gene Name
exocyst subunit exo70 family protein H1
Gene Aliases
AT3G55150, ATEXO70H1, exocyst subunit exo70 family protein H1
Chromosome Location
Chr.3: 20440491 - 20442650 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115373.4 NP_191075.2 1 - 1 1995 119
GenBank mRNA AY074262.1 - 1 - 1 1949 119

View Full Product Details