Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HEXO1
Entrez Gene ID
824692
Gene Name
beta-hexosaminidase 1
Gene Aliases
AT3G55260, ATHEX2, beta-hexosaminidase 1
Chromosome Location
Chr.3: 20489096 - 20493096 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115384.4 NP_567017.2 4 - 5 792 171
GenBank mRNA AM493720.1 - 4 - 5 571 171

View Full Product Details