Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBL
Entrez Gene ID
824716
Gene Name
Noc2p family
Gene Aliases
AT3G55510, REBELOTE
Chromosome Location
Chr.3: 20578922 - 20582505 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339730.1 NP_001326972.1 3 - 4 1215 65
NM_115408.4 NP_567023.1 4 - 5 1079 65
GenBank mRNA AK226780.1 - 4 - 5 1023 65

View Full Product Details