Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGP3
Entrez Gene ID
824770
Gene Name
UDP-glucose pyrophosphorylase 3
Gene Aliases
AT3G56040, UDP-glucose pyrophosphorylase 3
Chromosome Location
Chr.3: 20792477 - 20797670 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115462.4 NP_567031.1 4 - 5 1402 80

View Full Product Details