Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCH
Entrez Gene ID
824790
Gene Name
copper chaperone
Gene Aliases
AT3G56240, COPPER HOMEOSTASIS FACTOR, copper chaperone
Chromosome Location
Chr.3: 20863139 - 20864727 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339779.1 NP_001326129.1 2 - 3 594 83
NM_001339780.1 NP_001326128.1 3 - 4 275 83
NM_115482.4 NP_191183.1 2 - 3 301 83
GenBank mRNA AF361860.1 - 2 - 3 166 83
AY066056.1 - 1 - 2 81 83
AY085657.1 - 2 - 3 171 83
U88711.1 - 2 - 3 114 83

View Full Product Details