Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG18D
Entrez Gene ID
824811
Gene Name
autophagy 18 D-like protein
Gene Aliases
AT3G56440, ATATG18D, homolog of yeast autophagy 18 (ATG18) D
Chromosome Location
Chr.3: 20926001 - 20928993 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115502.4 NP_191203.2 4 - 5 1380 76
GenBank mRNA AK229064.1 - 4 - 5 1380 76

View Full Product Details