Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGL16
Entrez Gene ID
824890
Gene Name
AGAMOUS-like 16
Gene Aliases
AGAMOUS-like 16, AT3G57230
Chromosome Location
Chr.3: 21177282 - 21181026 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339845.1 NP_001319775.1 2 - 3 520 96
NM_001339847.1 NP_001326287.1 1 - 2 244 96
NM_115583.6 NP_191282.2 2 - 3 520 96
GenBank mRNA AY141209.1 - 1 - 2 183 96
BT030046.1 - 1 - 2 183 96

View Full Product Details