Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT3G57470
Entrez Gene ID
824914
Gene Name
Insulinase (Peptidase family M16) family protein
Gene Aliases
AT3G57470
Chromosome Location
Chr.3: 21269149 - 21276017 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203189.1 NP_001190118.1 20 - 21 2295 89
NM_001339871.1 NP_001319782.1 20 - 21 2207 89
NM_001339872.1 NP_001326452.1 20 - 21 2299 89
NM_001339873.1 NP_001326451.1 20 - 21 2299 89
NM_001339874.1 NP_001326450.1 20 - 21 2295 89
NM_115607.4 NP_567049.3 20 - 21 2203 89

View Full Product Details