The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035800.1 | NP_001030877.1 | 8 - 9 | 1272 | 107 |
NM_001339877.1 | NP_001325466.1 | 8 - 9 | 1089 | 107 | |
NM_115615.3 | NP_567051.1 | 8 - 9 | 1386 | 107 | |
GenBank mRNA | AF204676.1 | - | 8 - 9 | 1272 | 107 |
BT002466.1 | - | 8 - 9 | 1183 | 107 | |
BT009693.1 | - | 6 - 7 | 939 | 107 |