Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRPA1
Entrez Gene ID
824935
Gene Name
nuclear RNA polymerase A1
Gene Aliases
AT3G57660, nuclear RNA polymerase A1
Chromosome Location
Chr.3: 21353497 - 21363201 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339881.1 NP_001325828.1 17 - 18 3453 95
NM_115626.1 NP_191325.1 17 - 18 3243 95

View Full Product Details