Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS9B
Entrez Gene ID
830791
Gene Name
vacuolar protein sorting-associated 9A-like protein
Gene Aliases
AT5G09320, T5E8.120, T5E8_120
Chromosome Location
Chr.5: 2890102 - 2892094 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120968.3 NP_196494.2 2 - 3 311 100

View Full Product Details