Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT5G09315
Entrez Gene ID
28721089
Gene Name
vacuolar sorting protein 9 domain protein
Gene Aliases
AT5G09315
Chromosome Location
Chr.5: 2888659 - 2889953 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343044.1 NP_001330185.1 1 - 2 444 87
GenBank mRNA AY136390.1 - 1 - 2 371 87
BT006295.1 - 1 - 2 371 87

View Full Product Details