Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIZ1
Entrez Gene ID
836163
Gene Name
DNA-binding protein with MIZ/SP-RING zinc finger, PHD-finger and SAP domain-containing protein
Gene Aliases
AT5G60410, AT5G60420, ATSIZ1, MUF9.5, MUF9_5
Chromosome Location
Chr.5: 24294704 - 24301157 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037031.1 NP_001032108.1 9 - 10 908 80
NM_001037032.2 NP_001032109.2 9 - 10 908 80
NM_001125993.1 NP_001119465.1 9 - 10 908 80
NM_001345415.1 NP_001332003.1 9 - 10 938 80
NM_125434.4 NP_200849.2 9 - 10 1094 80
NM_203240.3 NP_974969.1 9 - 10 1037 80
GenBank mRNA AK175813.1 - 9 - 10 908 80
AK220973.1 - 9 - 10 908 80
AK221190.1 - 9 - 10 908 80
AK221249.1 - 9 - 10 908 80
AK221292.1 - 9 - 10 908 80
AK316983.1 - 9 - 10 908 80
AY139752.1 - 9 - 10 907 80
AY700572.1 - 9 - 10 909 80
BT004542.1 - 8 - 9 670 80

View Full Product Details