Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAMC2
Entrez Gene ID
840304
Gene Name
S-adenosylmethionine carrier 2
Gene Aliases
AT1G34065, S-adenosylmethionine carrier 2
Chromosome Location
Chr.1: 12398428 - 12401036 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333084.1 NP_001322412.1 3 - 4 209 85
NM_103129.5 NP_564436.4 3 - 4 230 85

View Full Product Details