Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB18
Entrez Gene ID
840987
Gene Name
RAB GTPASE HOMOLOG B18
Gene Aliases
ARABIDOPSIS RAB GTPASE HOMOLOG B18, ARABIDOPSIS RAB GTPASE HOMOLOG C1, AT1G43890, ATRAB-C1, ATRAB18, ATRABC1, F28H19.15, F28H19_15, RAB GTPASE HOMOLOG 18-1, RAB GTPASE HOMOLOG B18, RAB GTPASE HOMOLOG C1, RAB18-1, RABC1
Chromosome Location
Chr.1: 16646696 - 16648648 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084206.1 NP_001077675.1 4 - 5 388 85
NM_001123963.1 NP_001117435.1 3 - 4 394 85
NM_103516.4 NP_175056.1 4 - 5 424 85
GenBank mRNA AF370263.1 - 4 - 5 348 85
AK227664.1 - 4 - 5 383 85
AY063080.1 - 3 - 4 277 85
AY086878.1 - 4 - 5 387 85
U75603.1 - 3 - 4 296 85

View Full Product Details