Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCA3;4
Entrez Gene ID
841126
Gene Name
CYCLIN A3;4
Gene Aliases
AT1G47230, CYCLIN A3;4, F8G22.5, F8G22_5
Chromosome Location
Chr.1: 17306574 - 17308848 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103617.4 NP_175156.1 4 - 5 924 111
NM_202254.2 NP_973983.1 4 - 5 924 111
GenBank mRNA AK226253.1 - 4 - 5 930 111
AY087628.1 - 4 - 5 906 111

View Full Product Details