Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GA2OX4
- Entrez Gene ID
-
841217
- Gene Name
- gibberellin 2-oxidase 4
- Gene Aliases
-
AT1G47990, ATGA2OX4, Arabidopsis thaliana gibberellin 2-oxidase 4, T2J15.10, T2J15_10, gibberellin 2-oxidase 4
- Chromosome Location
-
Chr.1: 17698598 - 17701041 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103695.2 |
NP_175233.1 |
1 - 2 |
409 |
72 |
GenBank mRNA |
AY859740.1 |
- |
1 - 2 |
352 |
72 |
View Full Product Details