Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XIJ
Entrez Gene ID
824985
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
AT3G58160, ATMYOS3, ATXIJ, MYA3, MYOSIN A3, MYOSIN XI J, MYOSIN XI-16, XI-16
Chromosome Location
Chr.3: 21534064 - 21542127 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339909.1 NP_001319790.1 8 - 9 1295 75
NM_001339910.1 NP_001327763.1 8 - 9 1456 75
NM_001339911.1 NP_001327764.1 8 - 9 1456 75
NM_001339912.1 NP_001327762.1 8 - 9 1255 75
NM_115678.2 NP_191375.1 7 - 8 944 75

View Full Product Details