Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSY2
Entrez Gene ID
825044
Gene Name
citrate synthase 2
Gene Aliases
AT3G58750, citrate synthase 2
Chromosome Location
Chr.3: 21724306 - 21728137 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115737.3 NP_191434.1 11 - 12 2081 79
GenBank mRNA BT015884.1 - 11 - 12 1409 79
BT020195.1 - 11 - 12 1402 79

View Full Product Details