Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP65-4
Entrez Gene ID
825255
Gene Name
microtubule-associated protein 65-4
Gene Aliases
AT3G60840, microtubule-associated protein 65-4
Chromosome Location
Chr.3: 22477573 - 22480980 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340060.1 NP_001319808.1 12 - 13 1822 107

View Full Product Details