Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAUT1
Entrez Gene ID
825285
Gene Name
galacturonosyltransferase 1
Gene Aliases
AT3G61130, JS36, LGT1, galacturonosyltransferase 1
Chromosome Location
Chr.3: 22621912 - 22625808 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115977.4 NP_191672.1 9 - 10 1572 107
GenBank mRNA DQ370437.1 - 8 - 9 1170 107
KJ138782.1 - 8 - 9 1170 107

View Full Product Details