Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATCHX13
Entrez Gene ID
817574
Gene Name
Cation/hydrogen exchanger family protein
Gene Aliases
AT2G30240, CATION/H+ EXCHANGER 13, CHX13, T9D9.5, T9D9_5
Chromosome Location
Chr.2: 12899575 - 12902815 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128577.2 NP_180583.1 3 - 4 1697 64
GenBank mRNA EF571901.1 - 3 - 4 1661 64

View Full Product Details