Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMT1
Entrez Gene ID
831216
Gene Name
sterol methyltransferase 1
Gene Aliases
AT5G13710, CEPHALOPOD, CPH, MSH12.18, MSH12_18, SAM:CYCLOARTENOL-C24-METHYLTRANSFERASE, sterol methyltransferase 1
Chromosome Location
Chr.5: 4423555 - 4427324 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085110.1 NP_001078579.1 2 - 3 213 91
NM_121374.4 NP_196875.1 3 - 4 497 91
GenBank mRNA AF090372.1 - 3 - 4 164 91
AF195648.1 - 3 - 4 175 91
AY120716.1 - 3 - 4 189 91
BT000058.1 - 2 - 3 122 91

View Full Product Details